Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00944514_m1
          See other GET4 GE Assays ›
          Gene Symbol:
          GET4
          Gene Name:
          golgi to ER traffic protein 4
          Gene Aliases:
          C7orf20, CEE, CGI-20, TRC35
          Chromosome Location:
          Chr.7: 876554 - 896434 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00944514_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GET4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015949.2 NP_057033.2 3 - 4 412 72
          GenBank mRNA AF132954.1 - 3 - 4 299 72
          AK023560.1 - 3 - 4 336 72
          AK024305.1 - 2 - 3 1113 72
          AK097899.1 - 2 - 3 2073 72
          AK125863.1 - 2 - 3 2678 72
          AL133014.1 - 2 - 3 1202 72
          BC003550.1 - 3 - 4 354 3546669 72
          BM843540.1 - 3 - 4 412 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013324.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07003316.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GET4

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          cytoplasmic sequestering of protein
          tail-anchored membrane protein insertion into ER membrane
          maintenance of unfolded protein involved in ERAD pathway
          protein binding
          chaperone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000195.hg.1 TC07003316.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07000195.hg.1 ENST00000265857.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000195.hg.1 ENST00000407192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000195.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700196133.hg.1 TC0700013324.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000265857.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000407192.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000412734.6 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000426056.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000441491.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000464468.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 ENST00000483469.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Affymetrix Human Clariom D Assay TC0700013324.hg.1 NM_015949.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 156662 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline