| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01025558.hg.1 |
TC01001652.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01025558.hg.1 |
ENST00000304244.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01025558.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011152.hg.1 |
ENST00000304244.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181702.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181702.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181702.hg.1 |
TC0100011152.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100011152.hg.1 |
ENST00000304244.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011152.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181703.hg.1 |
ENST00000304244.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100181703.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181703.hg.1 |
ENST00000304244.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100181703.hg.1 |
NM_005298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100181703.hg.1 |
TC0100011152.hg.1 |
Clariom D TC |