| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000365536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011078.hg.1 |
ENST00000361822.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011078.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011078.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07011078.hg.1 |
TC07000707.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700162266.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162266.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162266.hg.1 |
TC0700008818.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000365536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000421864.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008818.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000361822.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000361946.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000425229.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000450657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
ENST00000457808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
NM_021994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700162265.hg.1 |
TC0700008818.hg.1 |
Clariom D TC |