Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057005.hg.1 |
TC01003712.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057005.hg.1 |
ENST00000367249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057005.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057005.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000473599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000478009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000483915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016973.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000367249.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000446185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000482572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
ENST00000497655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
NM_016243.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228586.hg.1 |
TC0100016973.hg.1 |
Clariom D TC |