Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00948450_m1
          See other NEB GE Assays ›
          Gene Symbol:
          NEB
          Gene Name:
          nebulin
          Gene Aliases:
          NEB177D, NEM2
          Chromosome Location:
          Chr.2: 151485334 - 151734487 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00948450_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164507.1 NP_001157979.1 44 - 45 5653 89
          NM_001164508.1 NP_001157980.1 44 - 45 5653 89
          NM_001271208.1 NP_001258137.1 44 - 45 5653 89
          NM_004543.4 NP_004534.2 44 - 45 5653 89
          XM_005246590.2 XP_005246647.1 44 - 45 5640 89
          XM_005246591.2 XP_005246648.1 44 - 45 5640 89
          XM_005246592.2 XP_005246649.1 44 - 45 5640 89
          XM_005246593.2 XP_005246650.1 44 - 45 5640 89
          XM_005246594.2 XP_005246651.1 44 - 45 5640 89
          XM_005246596.2 XP_005246653.1 44 - 45 5640 89
          XM_005246597.2 XP_005246654.1 44 - 45 5640 89
          XM_005246598.2 XP_005246655.1 44 - 45 5640 89
          XM_005246599.2 XP_005246656.1 44 - 45 5640 89
          XM_005246600.2 XP_005246657.1 44 - 45 5640 89
          XM_005246601.2 XP_005246658.1 44 - 45 5640 89
          XM_005246602.2 XP_005246659.1 44 - 45 5640 89
          XM_005246603.2 XP_005246660.1 44 - 45 5640 89
          XM_005246604.2 XP_005246661.1 44 - 45 5640 89
          XM_005246606.2 XP_005246663.1 44 - 45 5640 89
          XM_005246608.2 XP_005246665.1 44 - 45 5640 89
          XM_005246610.2 XP_005246667.1 44 - 45 5640 89
          XM_005246611.2 XP_005246668.1 44 - 45 5640 89
          XM_005246612.2 XP_005246669.1 44 - 45 5640 89
          XM_005246613.2 XP_005246670.1 44 - 45 5640 89
          XM_005246615.2 XP_005246672.1 44 - 45 5640 89
          XM_005246616.1 XP_005246673.1 44 - 45 5640 89
          XM_005246617.2 XP_005246674.1 44 - 45 5640 89
          XM_006712541.2 XP_006712604.1 44 - 45 5640 89
          XM_006712542.2 XP_006712605.1 44 - 45 5640 89
          XM_011511225.2 XP_011509527.1 44 - 45 5640 89
          XM_011511226.2 XP_011509528.1 44 - 45 5640 89
          XM_011511227.2 XP_011509529.1 44 - 45 5640 89
          XM_017004177.1 XP_016859666.1 44 - 45 5640 89
          XM_017004178.1 XP_016859667.1 44 - 45 5640 89
          XM_017004179.1 XP_016859668.1 44 - 45 5640 89
          XM_017004180.1 XP_016859669.1 44 - 45 5640 89
          XM_017004181.1 XP_016859670.1 44 - 45 5640 89
          XM_017004182.1 XP_016859671.1 44 - 45 5640 89
          XM_017004183.1 XP_016859672.1 44 - 45 5640 89
          XM_017004184.1 XP_016859673.1 44 - 45 5640 89
          XM_017004185.1 XP_016859674.1 44 - 45 5640 89
          GenBank mRNA X58122.1 - 8 - 9 1250 89
          X83957.1 - 31 - 32 5890 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014617.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002428.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NEB

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle organ development
          somatic muscle development
          muscle filament sliding
          regulation of actin filament length
          actin binding
          protein binding
          structural constituent of muscle

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 ENST00000172853.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 ENST00000397342.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 ENST00000397345.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 ENST00000409198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 ENST00000427231.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 NM_001164507.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 NM_001164508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 NM_004543.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02038245.hg.1 TC02002428.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000172853.14 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000397345.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000409198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000427231.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000603639.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000604864.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000618972.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001164507.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001164508.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001271208.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_004543.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000172853.14 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000397345.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000409198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000427231.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000603639.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000604864.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 ENST00000618972.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001164507.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001164508.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_001271208.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 NM_004543.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200206280.hg.1 TC0200014617.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline