| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006919.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006919.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006919.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006919.hg.1 |
TC03000324.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007450.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
ENST00000333127.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
ENST00000429548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
NM_203424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300153470.hg.1 |
TC0300007450.hg.1 |
Clariom D TC |