| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029425.hg.1 |
TC05002120.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029425.hg.1 |
ENST00000274606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029425.hg.1 |
ENST00000314397.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029425.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029425.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029423.hg.1 |
TC05002120.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029423.hg.1 |
ENST00000274606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05029423.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190953.hg.1 |
TC0500013010.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500190951.hg.1 |
TC0500013010.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000274606.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000314397.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000502263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000510363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000511078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
ENST00000514354.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_001034833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013010.hg.1 |
NM_017838.3 |
Refseq |