Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00951825_g1
          See other FAM183BP GE Assays ›
          Gene Symbol:
          FAM183BP
          Gene Name:
          acyloxyacyl hydrolase (neutrophil)
          Gene Aliases:
          FAM183B, THEG6
          Chromosome Location:
          Chr.7: 38685346 - 38687089 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00951825_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAM183BP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028347.1 1 - 2 773 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010792.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001304.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05430264 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P391991 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P391991 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17222 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 54442 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P391991 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P391991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15931674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651577 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430333 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15931674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651577 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651582 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25651571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05430333 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P391991 - -
          Agilent Human 1B Microarray G4111A A_32_P27903 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8139164 - -
          Affymetrix Human Gene 1.0 ST Array 8139161 - -
          Affymetrix Human Gene 1.0 ST Array 8139162 - -
          Affymetrix Human Gene 1.1 ST Array 8139160 - -
          Affymetrix Human Gene 1.1 ST Array 8139163 - -
          Affymetrix Human Genome U219 Array 11735672_PM_a_at - -
          Affymetrix Human Genome U219 Array 11735672_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR07020229.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020229.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020229.hg.1 TC07001304.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700010792.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010792.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020230.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020230.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020230.hg.1 TC07001304.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07020231.hg.1 NR_028347.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020231.hg.1 TC07001304.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0700010792.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010792.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010792.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176868.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176868.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176868.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176868.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176868.hg.1 TC0700010792.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700176870.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176870.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176870.hg.1 TC0700010792.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700176869.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176869.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176869.hg.1 ENST00000409072.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0700176869.hg.1 NR_028347.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700176869.hg.1 TC0700010792.hg.1 Clariom D TC
          Human Genome U95 Set 51959_PM_at - -
          Human Genome U95 Set 51959_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032704 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032704 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 100727 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3244033 BC045803.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3244033 NR_028347.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3244033 AI217903.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3244033 AK124132.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244033 BC045803.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244033 NR_028347.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244033 AI217903.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3244033 AK124132.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline