| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02023363.hg.1 |
ENST00000305959.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02023363.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02023363.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02023363.hg.1 |
TC02001450.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200181440.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200181440.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200181440.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200181440.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200181440.hg.1 |
TC0200011274.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
ENST00000409223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011274.hg.1 |
NM_198582.3 |
Refseq |