Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00956167_s1
          See other FAM133CP GE Assays ›
          Gene Symbol:
          FAM133CP
          Gene Name:
          family with sequence similarity 133, member A pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 58715015 - 58717534 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00956167_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAM133CP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027508.1 1 464 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000007708.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000363.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Oligo Microarray 4x44K 23563 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P247536 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P233938 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P158065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631358 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631368 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631378 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631368 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249127 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631358 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27631350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07249098 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249097 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249122 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249102 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249100 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249130 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249086 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249124 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249084 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249107 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249103 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07249109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16346362 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8140951 - -
          Affymetrix Human Gene 1.0 ST Array 8140948 - -
          Affymetrix Human Gene 1.0 ST Array 8105505 - -
          Affymetrix Human Gene 1.0 ST Array 8140944 - -
          Affymetrix Human Gene 1.0 ST Array 8140954 - -
          Affymetrix Human Gene 1.0 ST Array 8140952 - -
          Affymetrix Human Gene 1.0 ST Array 8055979 - -
          Affymetrix Human Gene 1.0 ST Array 8140946 - -
          Affymetrix Human Gene 1.0 ST Array 8140949 - -
          Affymetrix Human Gene 1.0 ST Array 8140950 - -
          Affymetrix Human Gene 1.1 ST Array 8055978 - -
          Affymetrix Human Gene 1.1 ST Array 8140942 - -
          Affymetrix Human Transcriptome Array 2.0 TC1200010167.hg.1 ENST00000538746.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007708.hg.1 ENST00000332869.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10004207.hg.1 NR_027508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10004207.hg.1 TC10000363.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000007708.hg.1 ENST00000332869.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay TC1000007708.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152392.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152392.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152392.hg.1 TC1000007708.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010167.hg.1 ENST00000538746.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000152393.hg.1 ENST00000332869.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000152393.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152393.hg.1 ENST00000332869.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000152393.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152393.hg.1 TC1000007708.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000152394.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152394.hg.1 NR_027508.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000152394.hg.1 TC1000007708.hg.1 Clariom D TC
          Human Genome U95 Set 89639_at - -
          Human Genome U95 Set 89639_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043978 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043978 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 101123 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3225406 NR_027508.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3225406 AA447601.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline