| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005254.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005254.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005254.hg.1 |
TC20000363.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005253.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005253.hg.1 |
ENST00000545925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005253.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005253.hg.1 |
TC20000363.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005251.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005251.hg.1 |
ENST00000545925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005251.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005251.hg.1 |
TC20000363.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005252.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005252.hg.1 |
ENST00000545925.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005252.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005252.hg.1 |
TC20000363.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000152448.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000152448.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000152448.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000152448.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000152448.hg.1 |
TC2000007514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000152447.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000152447.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000152447.hg.1 |
ENST00000372330.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000152447.hg.1 |
NM_004994.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000152447.hg.1 |
TC2000007514.hg.1 |
Clariom D TC |