Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00961183_m1
          See other TNKS2 GE Assays ›
          Gene Symbol:
          TNKS2
          Gene Name:
          tankyrase 2
          Gene Aliases:
          ARTD6, PARP-5b, PARP-5c, PARP5B, PARP5C, TANK2, TNKL, pART6
          Chromosome Location:
          Chr.10: 91798426 - 91865475 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00961183_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNKS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025235.4 NP_079511.1 11 - 12 1545 104
          XM_011540213.2 XP_011538515.1 11 - 12 1608 104
          XM_017016699.2 XP_016872188.1 10 - 11 1097 104
          XM_017016701.2 XP_016872190.1 11 - 12 1608 104
          XM_047425795.1 XP_047281751.1 11 - 12 1545 104
          GenBank mRNA AF264912.1 - 11 - 12 1280 104
          AF305081.1 - 11 - 12 1579 104
          AF309033.1 - 11 - 12 1531 104
          AF329696.1 - 11 - 12 1419 104
          AF342982.1 - 11 - 12 1280 104
          AF438201.1 - 11 - 12 1659 104
          AK314612.1 - 11 - 12 1545 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008447.hg
          This assay targets the same location as HTA transcript cluster(s) TC10000654.hg

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNKS2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          Wnt signaling pathway
          positive regulation of telomere maintenance via telomerase
          protein localization to chromosome, telomeric region
          protein poly-ADP-ribosylation
          protein auto-ADP-ribosylation
          positive regulation of canonical Wnt signaling pathway
          positive regulation of telomere capping
          negative regulation of telomere maintenance via telomere lengthening
          NAD+ ADP-ribosyltransferase activity
          protein binding
          transferase activity
          transferase activity, transferring glycosyl groups
          nucleotidyltransferase activity
          enzyme binding
          metal ion binding
          NAD+-protein-aspartate ADP-ribosyltransferase activity
          NAD+-protein-glutamate ADP-ribosyltransferase activity
          protein ADP-ribosylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10007275.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10007275.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10007275.hg.1 TC10000654.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157366.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157366.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157366.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000157366.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000157366.hg.1 TC1000008447.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 ENST00000371627.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq
          Affymetrix Human Clariom D Assay TC1000008447.hg.1 NM_025235.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0