Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00961458_m1
          See other CD2AP GE Assays ›
          Gene Symbol:
          CD2AP
          Gene Name:
          CD2 associated protein
          Gene Aliases:
          CMS
          Chromosome Location:
          Chr.6: 47477789 - 47627263 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00961458_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD2AP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012120.3 NP_036252.1 6 - 7 1183 80
          XM_005248976.2 XP_005249033.1 6 - 7 1183 80
          XM_011514449.3 XP_011512751.1 5 - 6 693 80
          XM_017010641.2 XP_016866130.1 6 - 7 1183 80
          GenBank mRNA AF146277.1 - 6 - 7 777 80
          AF164377.1 - 5 - 6 727 80
          BC069444.1 - 6 - 7 773 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600008183.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000633.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD2AP

          Gene Ontology Categories:

          Function(s) Process(es)

          immunological synapse formation
          kidney development
          liver development
          endothelium development
          lipid metabolic process
          apoptotic process
          substrate-dependent cell migration, cell extension
          inflammatory response
          response to oxidative stress
          actin filament organization
          signal transduction
          cell proliferation
          male gonad development
          protein secretion
          response to wounding
          response to virus
          gene expression
          protein transport
          cell migration
          actin filament polymerization
          cell differentiation
          protein catabolic process
          neuron projection development
          Rab protein signal transduction
          glomerulus development
          response to insulin
          transforming growth factor beta1 production
          negative regulation of transforming growth factor beta1 production
          regulation of actin cytoskeleton organization
          maintenance of blood-brain barrier
          neurotrophin signaling pathway
          nerve growth factor signaling pathway
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          cell-cell adhesion mediated by cadherin
          cell-cell junction organization
          glucose import
          filopodium assembly
          neurotrophin TRK receptor signaling pathway
          regulation of synaptic plasticity
          lymph node development
          collateral sprouting
          positive regulation of protein secretion
          synapse organization
          T cell receptor signaling pathway
          negative regulation of small GTPase mediated signal transduction
          protein heterooligomerization
          cell division
          stress-activated MAPK cascade
          localization of cell
          adipose tissue development
          membrane organization
          protein-containing complex assembly
          ERK1 and ERK2 cascade
          response to transforming growth factor beta
          glomerular visceral epithelial cell differentiation
          renal albumin absorption
          positive regulation of protein localization to nucleus
          response to glial cell derived neurotrophic factor
          actin binding
          structural constituent of cytoskeleton
          protein binding
          SH3 domain binding
          clathrin binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          identical protein binding
          cadherin binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000359314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000463175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000463175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000463175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000463175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000477159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000477159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000477159.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000479857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000479857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000479857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000479857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000486693.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000486693.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000486693.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 ENST00000486693.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008183.hg.1 NM_012120.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline