Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00961709_m1
          See other CLDN19 GE Assays ›
          Gene Symbol:
          CLDN19
          Gene Name:
          claudin 19
          Gene Aliases:
          HOMG5
          Chromosome Location:
          Chr.1: 42733093 - 42740254 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00961709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLDN19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001123395.1 NP_001116867.1 1 - 2 416 63
          NM_001185117.1 NP_001172046.1 1 - 2 416 63
          NM_148960.2 NP_683763.2 1 - 2 416 63
          GenBank mRNA AF497644.1 - 1 - 2 225 63
          AK096063.1 - 1 - 2 416 63
          AK291197.1 - 1 - 2 416 63
          AK298992.1 - 1 - 2 395 63
          BC030524.1 - 1 - 2 302 5207628 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013896.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002573.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLDN19

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          neuronal action potential propagation
          apical junction assembly
          response to stimulus
          structural molecule activity
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 TC01002573.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 ENST00000296387.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 ENST00000539749.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039978.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 TC01002573.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 ENST00000296387.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 ENST00000539749.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039976.hg.1 NM_148960.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 TC01002573.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 ENST00000296387.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 ENST00000539749.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01039977.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203845.hg.1 TC0100013896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203846.hg.1 TC0100013896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203847.hg.1 TC0100013896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100203848.hg.1 TC0100013896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000296387.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000372539.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 ENST00000539749.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001123395.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_001185117.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013896.hg.1 NM_148960.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline