| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034116.hg.1 |
TC01002207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034116.hg.1 |
ENST00000376572.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034116.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034116.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034115.hg.1 |
TC01002207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034115.hg.1 |
ENST00000376572.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034115.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034115.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009506.hg.1 |
ENST00000567634.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195359.hg.1 |
TC0100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195358.hg.1 |
TC0100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600009506.hg.1 |
ENST00000567634.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195361.hg.1 |
TC0100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012901.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195362.hg.1 |
TC0100012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000376572.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000376576.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
ENST00000616327.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
NM_138346.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195360.hg.1 |
TC0100012901.hg.1 |
Clariom D TC |