Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00962350_m1
          See other SCN1B GE Assays ›
          Gene Symbol:
          SCN1B
          Gene Name:
          sodium voltage-gated channel beta subunit 1
          Gene Aliases:
          ATFB13, BRGDA5, GEFSP1
          Chromosome Location:
          Chr.19: 35030688 - 35040449 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00962350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCN1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037.4 NP_001028.1 2 - 3 345 59
          NM_001321605.1 NP_001308534.1 2 - 3 585 59
          NM_199037.4 NP_950238.1 2 - 3 345 59
          GenBank mRNA AK313279.1 - 2 - 3 230 59
          AY391842.1 - 2 - 3 212 59
          BC021266.2 - 1 - 2 116 59
          BC067122.1 - 2 - 3 345 5728955 59
          BM805468.1 - 2 - 3 367 59
          BT019923.1 - 2 - 3 212 59
          DA062026.1 - 2 - 3 221 59
          L10338.1 - 2 - 3 309 59
          L16242.1 - 2 - 3 236 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007830.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000455.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Recommended Assay Homo sapiens Human Cholera Infection Inventoried Human_Cholera_Infection Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SCN1B

          Panther Classification:

          Molecular Function -

          ion channel transporter voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          chemical synaptic transmission
          axon guidance
          positive regulation of sodium ion transport
          positive regulation of neuron projection development
          neuronal action potential propagation
          corticospinal neuron axon guidance
          sodium ion transmembrane transport
          locomotion
          response to pyrethroid
          membrane depolarization
          cardiac muscle contraction
          regulation of postsynaptic membrane potential
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          cardiac conduction
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during Purkinje myocyte cell action potential
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transporter activity
          voltage-gated sodium channel activity
          protein binding
          sodium channel regulator activity
          sodium channel inhibitor activity
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential
          voltage-gated sodium channel activity involved in Purkinje myocyte action potential

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19005955.hg.1 ENST00000415950.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005955.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005955.hg.1 TC19000455.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000601692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000601692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900157716.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157716.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900157716.hg.1 TC1900007830.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157716.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157716.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900157715.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157715.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900157715.hg.1 TC1900007830.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157715.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157715.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000262631.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000415950.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000595652.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000596348.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000601692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000601692.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 ENST00000602150.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_001037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq
          Affymetrix Human Clariom D Assay TC1900007830.hg.1 NM_199037.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline