Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00962356_m1
          See other ZC3H12A GE Assays ›
          Gene Symbol:
          ZC3H12A
          Gene Name:
          zinc finger CCCH-type containing 12A
          Gene Aliases:
          MCPIP, MCPIP-1, MCPIP1, Reg1, dJ423B22.1
          Chromosome Location:
          Chr.1: 37474580 - 37484377 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00962356_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZC3H12A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001323550.2 NP_001310479.1 3 - 4 671 95
          NM_001323551.2 NP_001310480.1 3 - 4 674 95
          NM_025079.3 NP_079355.2 3 - 4 674 95
          XM_011542198.3 XP_011540500.1 3 - 4 674 95
          XM_047430884.1 XP_047286840.1 2 - 3 918 95
          XM_047430885.1 XP_047286841.1 3 - 4 671 95
          XM_047430886.1 XP_047286842.1 3 - 4 671 95
          GenBank mRNA AK026884.1 - 3 - 4 702 95
          AK093343.1 - 3 - 4 528 95
          AY920403.1 - 2 - 3 586 95
          BC005001.1 - 3 - 4 660 95
          CR457364.1 - 2 - 3 586 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007832.hg
          This assay targets the same location as HTA transcript cluster(s) TC01000474.hg

          Additional Information:

          This assay is in the 5'UTR region of the NM_001323551.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZC3H12A

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          angiogenesis
          negative regulation of protein phosphorylation
          positive regulation of defense response to virus by host
          immune system process
          immune response-activating signal transduction
          mRNA catabolic process
          apoptotic process
          inflammatory response
          cellular response to DNA damage stimulus
          nervous system development
          regulation of gene expression
          positive regulation of autophagy
          miRNA catabolic process
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of lipid storage
          protein deubiquitination
          cell differentiation
          negative regulation of interferon-gamma production
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          cellular response to oxidative stress
          cellular response to glucose starvation
          positive regulation of protein import into nucleus
          negative regulation of macrophage activation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation by host of viral genome replication
          negative regulation of nitric oxide biosynthetic process
          positive regulation of fat cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          T cell receptor signaling pathway
          protein oligomerization
          defense response to virus
          negative regulation of cardiac muscle contraction
          positive regulation of mRNA catabolic process
          3'-UTR-mediated mRNA destabilization
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to virus
          negative regulation of cytokine production involved in inflammatory response
          positive regulation of execution phase of apoptosis
          positive regulation of p38MAPK cascade
          negative regulation of NIK/NF-kappaB signaling
          positive regulation of protein deubiquitination
          cellular response to sodium arsenite
          cellular response to ionomycin
          cellular response to chemokine
          negative regulation of T-helper 17 cell differentiation
          positive regulation of reactive oxygen species metabolic process
          positive regulation of miRNA catabolic process
          DNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          nuclease activity
          endonuclease activity
          endoribonuclease activity
          exoribonuclease activity
          ribonuclease activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          zinc ion binding
          hydrolase activity
          miRNA binding
          RNA stem-loop binding
          mRNA 3'-UTR AU-rich region binding
          ribosome binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 ENST00000619365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007832.hg.1 NR_106790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007358.hg.1 ENST00000373082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007358.hg.1 ENST00000373087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007358.hg.1 ENST00000492829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007358.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007358.hg.1 TC01000474.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01007359.hg.1 ENST00000373082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007359.hg.1 ENST00000373087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007359.hg.1 ENST00000492829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007359.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007359.hg.1 TC01000474.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01007361.hg.1 ENST00000373082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007361.hg.1 ENST00000373087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007361.hg.1 ENST00000492829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007361.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007361.hg.1 TC01000474.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01007360.hg.1 ENST00000373082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007360.hg.1 ENST00000373087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007360.hg.1 ENST00000492829.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007360.hg.1 NM_025079.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007360.hg.1 TC01000474.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 ENST00000619365.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007832.hg.1 NR_106790.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155140.hg.1 TC0100007832.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155138.hg.1 TC0100007832.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155137.hg.1 TC0100007832.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000373087.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000471012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 ENST00000472312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 NM_025079.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100155139.hg.1 TC0100007832.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline