| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046986.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046986.hg.1 |
ENST00000391989.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046986.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046986.hg.1 |
TC02002934.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016309.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
ENST00000319460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
ENST00000391989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
NM_148961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200219395.hg.1 |
TC0200016309.hg.1 |
Clariom D TC |