| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001733.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001733.hg.1 |
ENST00000377777.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001733.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001733.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001733.hg.1 |
TC06000169.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007274.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001734.hg.1 |
ENST00000377777.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001734.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001734.hg.1 |
TC06000169.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600149336.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149336.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149336.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149336.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149336.hg.1 |
TC0600007274.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007274.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
ENST00000289316.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
ENST00000377777.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
NM_021063.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
NM_138720.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600149334.hg.1 |
TC0600007274.hg.1 |
Clariom D TC |