| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010628.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010628.hg.1 |
ENST00000465017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010628.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010628.hg.1 |
TC09002882.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012189.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000290079.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000465017.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000479737.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000483187.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000484854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
ENST00000489739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
NM_032928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900185227.hg.1 |
TC0900012189.hg.1 |
Clariom D TC |