| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004873.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004873.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004873.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004873.hg.1 |
TC06000415.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001919.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001919.hg.1 |
TC6_dbb_hap3000097.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001547.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001547.hg.1 |
TC6_mann_hap4000089.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001714.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001714.hg.1 |
TC6_mcf_hap5000090.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001935.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001935.hg.1 |
TC6_cox_hap2000105.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001918.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001918.hg.1 |
TC6_dbb_hap3000097.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004872.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004872.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004872.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004872.hg.1 |
TC06000415.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001546.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001546.hg.1 |
TC6_mann_hap4000089.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001899.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001899.hg.1 |
TC6_qbl_hap6000096.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001900.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001900.hg.1 |
TC6_qbl_hap6000096.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001936.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001936.hg.1 |
TC6_cox_hap2000105.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001713.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001713.hg.1 |
TC6_mcf_hap5000090.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600153138.hg.1 |
TC0600007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000374656.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
ENST00000478431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007687.hg.1 |
NM_002931.3 |
Refseq |