Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00969366_m1
          See other MBD2 GE Assays ›
          Gene Symbol:
          MBD2
          Gene Name:
          methyl-CpG binding domain protein 2
          Gene Aliases:
          DMTase, NY-CO-41
          Chromosome Location:
          Chr.18: 54151606 - 54224669 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00969366_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MBD2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003927.5 NP_003918.1 1 - 2 652 62
          NM_015832.6 NP_056647.1 1 - 2 652 62
          GenBank mRNA AF072242.1 - 1 - 2 771 62
          AF072246.1 - 1 - 2 74 62
          BC032638.1 - 1 - 2 612 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800008730.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000521.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human DNA Methylation and Transcriptional Repression Inventoried Human_DNA_Methylation_and_Transcriptional_Repression Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MBD2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ventricular cardiac muscle tissue development
          chromatin remodeling
          methylation-dependent chromatin silencing
          regulation of transcription from RNA polymerase II promoter
          response to mechanical stimulus
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          response to nutrient levels
          response to estradiol
          regulation of gene expression, epigenetic
          regulation of cell proliferation
          maternal behavior
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          embryonic organ development
          protein-containing complex assembly
          response to bisphenol A
          cellular response to serotonin
          DNA binding
          chromatin binding
          satellite DNA binding
          mRNA binding
          protein binding
          methyl-CpG binding
          protein domain specific binding
          nucleosomal DNA binding
          siRNA binding
          identical protein binding
          binding, bridging
          C2H2 zinc finger domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005427.hg.1 ENST00000256429.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005427.hg.1 ENST00000398398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005427.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005427.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005427.hg.1 TC18000521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18005426.hg.1 ENST00000256429.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005426.hg.1 ENST00000398398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005426.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005426.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005426.hg.1 TC18000521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18005428.hg.1 ENST00000256429.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005428.hg.1 ENST00000398398.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18005428.hg.1 NM_003927.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005428.hg.1 NM_015832.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18005428.hg.1 TC18000521.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159874.hg.1 TC1800008730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159879.hg.1 TC1800008730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159875.hg.1 TC1800008730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000578272.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000579025.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay TC1800008730.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159878.hg.1 TC1800008730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000256429.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000398398.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 ENST00000583046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 NM_003927.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 NM_015832.4 Refseq
          Affymetrix Human Clariom D Assay PSR1800159876.hg.1 TC1800008730.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0