| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003367.hg.1 |
TC05000273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003367.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003367.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003373.hg.1 |
TC05000273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003373.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003373.hg.1 |
ENST00000512767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003373.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003365.hg.1 |
TC05000273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003365.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003365.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003366.hg.1 |
TC05000273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003366.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003366.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007590.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151185.hg.1 |
TC0500007590.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000512767.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
ENST00000513930.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151193.hg.1 |
TC0500007590.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500151184.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151184.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151184.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151184.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151184.hg.1 |
TC0500007590.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151187.hg.1 |
TC0500007590.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000389074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000506473.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
ENST00000509189.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
NM_001164442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151186.hg.1 |
TC0500007590.hg.1 |
Clariom D TC |