Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00971643_g1
          See other SRD5A1 GE Assays ›
          Gene Symbol:
          SRD5A1
          Gene Name:
          steroid 5 alpha-reductase 1
          Gene Aliases:
          S5AR 1
          Chromosome Location:
          Chr.5: 6633440 - 6674386 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00971643_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SRD5A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047.4 NP_001038.1 1 - 2 429 75
          NR_136739.2 1 - 2 429 75
          GenBank mRNA AF052126.1 - 1 - 2 419 75
          AF113128.1 - 2 - 3 306 75
          AK313070.1 - 1 - 2 429 75
          BC006373.2 - 1 - 2 400 75
          BC007033.1 - 1 - 2 429 75
          BC008673.2 - 1 - 2 403 75
          BT006834.1 - 1 - 2 292 75
          DA170063.1 - 1 - 2 502 75
          M32313.1 - 1 - 2 322 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500013149.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000044.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SRD5A1

          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          liver development
          lipid metabolic process
          steroid biosynthetic process
          androgen biosynthetic process
          androgen catabolic process
          sex determination
          sex differentiation
          steroid metabolic process
          androgen metabolic process
          male gonad development
          cellular response to starvation
          response to xenobiotic stimulus
          response to muscle activity
          diterpenoid metabolic process
          spinal cord development
          hippocampus development
          thalamus development
          hypothalamus development
          pituitary gland development
          cerebral cortex development
          cell differentiation
          male genitalia development
          female genitalia development
          response to follicle-stimulating hormone
          response to estradiol
          cellular response to insulin stimulus
          response to testosterone
          serotonin metabolic process
          progesterone metabolic process
          response to estrogen
          bone development
          response to growth hormone
          response to fungicide
          cellular response to cAMP
          cellular response to growth factor stimulus
          cellular response to estradiol stimulus
          cellular response to testosterone stimulus
          cellular response to dexamethasone stimulus
          cellular response to epinephrine stimulus
          cellular response to serotonin
          3-oxo-5-alpha-steroid 4-dehydrogenase activity
          protein binding
          electron carrier activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors
          amide binding
          cholestenone 5-alpha-reductase activity
          NADPH binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05000519.hg.1 ENST00000274192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000519.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000519.hg.1 TC05000044.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00008577.hg.1 ENST00000442297.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 ENST00000274192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 ENST00000537411.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 ENST00000538824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000524.hg.1 TC05000044.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05000526.hg.1 ENST00000274192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000526.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000526.hg.1 ENST00000537411.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000526.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000526.hg.1 TC05000044.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192667.hg.1 TC0500013149.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192669.hg.1 TC0500013149.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500192662.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192662.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192662.hg.1 TC0500013149.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00008577.hg.1 ENST00000442297.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008577.hg.1 NR_028597.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192666.hg.1 TC0500013149.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline