Search
Search

OR2L2| Gene Symbol: OR2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004686.2 | NP_001004686.1 | 1 | 247 | 133 | |
| XM_011544158.2 | XP_011542460.1 | 2 | 377 | 133 | ||
| GenBank mRNA | BC104792.1 | - | 1 | 247 | 8143795 | 133 |
| BC104794.1 | - | 1 | 247 | 8143797 | 133 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005492 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005499 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931264 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369979 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0417 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0418 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29223 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369979 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3369979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005499 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005492 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005499 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931261 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931280 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931258 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931270 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931256 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931276 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001004686_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7911254 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7911253 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7911255 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7911252 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1567238_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1567240_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1567237_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735953_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735953_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031113.hg.1 | TC01002026.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031113.hg.1 | ENST00000366479.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031113.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190602.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190602.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190602.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190602.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190602.hg.1 | TC0100012343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100190604.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190604.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190604.hg.1 | TC0100012343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012343.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190603.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190603.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190603.hg.1 | ENST00000366479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190603.hg.1 | NM_001004686.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190603.hg.1 | TC0100012343.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1567237_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567240_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567240_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567238_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567238_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1567237_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001004686_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567240_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567238_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1567237_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042315 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 191139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1680225 | X64978.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_138871-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5820133 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_138871 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1680225 | NM_001004686.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680225 | BC104792.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680225 | BC104794.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680225 | X64978.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680225 | NM_001004686.2 | human_transcriptome |