Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00975302_g1
          See other SLAMF8 GE Assays ›
          Gene Symbol:
          SLAMF8
          Gene Name:
          SLAM family member 8
          Gene Aliases:
          BLAME, CD353, SBBI42
          Chromosome Location:
          Chr.1: 159826873 - 159837492 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00975302_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLAMF8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001330741.2 NP_001317670.1 2 - 3 376 65
          NM_020125.3 NP_064510.1 3 - 4 703 65
          XM_005245345.4 XP_005245402.1 3 - 4 703 65
          XM_011509775.3 XP_011508077.1 3 - 4 703 65
          XM_011509776.3 XP_011508078.1 3 - 4 685 65
          XM_011509777.4 XP_011508079.1 3 - 4 703 65
          XM_011509778.3 XP_011508080.1 3 - 4 574 65
          XM_011509779.3 XP_011508081.1 2 - 3 376 65
          XM_017001844.3 XP_016857333.1 3 - 4 703 65
          XM_017001845.3 XP_016857334.1 3 - 4 703 65
          XM_017001846.2 XP_016857335.1 3 - 4 685 65
          GenBank mRNA AF146761.1 - 3 - 4 765 65
          AK074669.1 - 3 - 4 826 65
          BC109194.2 - 3 - 4 703 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010269.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001357.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLAMF8

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte chemotaxis involved in inflammatory response
          B-1 B cell lineage commitment
          immune response
          negative regulation of macrophage chemotaxis
          defense response to bacterium
          regulation of B cell differentiation
          negative regulation of respiratory burst involved in inflammatory response
          negative regulation of monocyte chemotaxis
          phagosome acidification
          negative regulation of neutrophil migration
          negative regulation of dendritic cell chemotaxis
          protein binding
          signaling receptor activity
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01021260.hg.1 ENST00000289707.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021260.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021260.hg.1 ENST00000471286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01021260.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01021260.hg.1 TC01001357.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010269.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000368102.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000491974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 ENST00000621242.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018513.hg.1 NM_001134233.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000289707.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000368104.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000471286.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 ENST00000497141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 NM_020125.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100175337.hg.1 TC0100010269.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline