| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
ENST00000370022.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
ENST00000370025.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
ENST00000467302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014634.hg.1 |
TC01000932.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
ENST00000485810.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009328.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370022.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000370025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
ENST00000467302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
NM_018061.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
NR_037185.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166429.hg.1 |
TC0100009328.hg.1 |
Clariom D TC |