| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000220876.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
ENST00000518491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_001199214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008044.hg.1 |
NM_007029.3 |
Refseq |