Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00976691_m1
          See other WTIP GE Assays ›
          Gene Symbol:
          WTIP
          Gene Name:
          Wilms tumor 1 interacting protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 34481935 - 34509147 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00976691_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WTIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080436.1 NP_001073905.1 5 - 6 1028 70
          XM_006723014.3 XP_006723077.1 5 - 6 1061 70
          XM_011526452.2 XP_011524754.1 5 - 6 1068 70
          XM_011526453.2 XP_011524755.1 5 - 6 462 70
          GenBank mRNA AK130059.1 - 4 - 5 366 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007810.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000445.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cytoskeleton organization
          regulation of cell morphogenesis
          gene silencing by miRNA
          negative regulation of hippo signaling
          negative regulation of nucleic acid-templated transcription
          positive regulation of gene silencing by miRNA
          transcription corepressor activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19005824.hg.1 ENST00000270288.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005824.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005824.hg.1 TC19000445.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 TC1900007810.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157492.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000585928.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 ENST00000590071.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007810.hg.1 NM_001080436.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline