| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024329.hg.1 |
TC01001577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024329.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024329.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179644.hg.1 |
TC0100010843.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000367563.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
ENST00000612603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010843.hg.1 |
NR_034131.1 |
Refseq |