Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00977097_s1
          See other PRO2964 GE Assays ›
          Gene Symbol:
          PRO2964
          Gene Name:
          uncharacterized protein PRO2964
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00977097_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRO2964
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF119910.1 - 1 863 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream proline-rich protein 18-like LOC105373780 -5.2kb Sense Chr. 2 - 184593627 - 184598985 on NCBI Build GRCh38
          Downstream uncharacterized LOC105373782 LOC105373782 406.1kb Sense Chr. 2 - 185164954 - 185167072 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P430156 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4712 - -
          SurePrint G3 Human GE Microarray G4851A ERCC-00033_60 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P430156 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P430156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8057559 - -
          Affymetrix HT HG-U133+ PM Array Plate 220872_PM_at - -
          Human Genome U133 Plus 2.0 Array 220872_at - -
          Human Genome U133 Plus 2.0 Array 220872_PM_at - -
          Human Genome U133A 2.0 Array 220872_at - -
          Human Genome U133A 2.0 Array 220872_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw502957 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220872_at1 - -
          Human Genome U133 Set 220872_PM_at - -
          Human Genome U133 Set 220872_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0