Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00977600_g1
          See other CHMP6 GE Assays ›
          Gene Symbol:
          CHMP6
          Gene Name:
          charged multivesicular body protein 6
          Gene Aliases:
          VPS20
          Chromosome Location:
          Chr.17: 80991841 - 81000133 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00977600_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHMP6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024591.5 NP_078867.2 3 - 4 336 84
          XM_005257668.1 XP_005257725.1 3 - 4 336 84
          GenBank mRNA AK021811.1 - 3 - 4 274 84
          AK223187.1 - 3 - 4 298 84
          AK292105.1 - 3 - 4 302 84
          AY329087.1 - 3 - 4 258 84
          BC010108.2 - 3 - 4 282 84
          BP196406.1 - 3 - 4 336 84
          CR457284.1 - 3 - 4 258 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009144.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000922.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHMP6

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          membrane budding
          autophagy
          nucleus organization
          vacuolar transport
          mitotic metaphase plate congression
          negative regulation of epidermal growth factor-activated receptor activity
          protein transport
          macroautophagy
          nuclear envelope reassembly
          late endosome to vacuole transport via multivesicular body sorting pathway
          multivesicular body assembly
          viral budding via host ESCRT complex
          regulation of protein catabolic process
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          viral budding from plasma membrane
          vesicle fusion with vacuole
          multivesicular body-lysosome fusion
          midbody abscission
          multivesicular body sorting pathway
          membrane fission
          autophagosome maturation
          regulation of mitotic spindle assembly
          late endosome to lysosome transport
          regulation of exosomal secretion
          ESCRT III complex assembly
          protein binding
          macromolecular complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17012135.hg.1 ENST00000325167.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012135.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012135.hg.1 TC17000922.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012134.hg.1 ENST00000325167.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17012134.hg.1 NM_024591.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17012134.hg.1 TC17000922.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009144.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700169293.hg.1 TC1700009144.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000325167.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000571457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000572525.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 ENST00000572778.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 NM_024591.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700169294.hg.1 TC1700009144.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline