| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030899.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030899.hg.1 |
ENST00000383659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030899.hg.1 |
ENST00000465044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030899.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030899.hg.1 |
TC03001706.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000463266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000471534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
ENST00000478601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012195.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000306103.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000465044.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
ENST00000467643.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
NM_024610.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190884.hg.1 |
TC0300012195.hg.1 |
Clariom D TC |