Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00983296_s1
          See other LOC1720 GE Assays ›
          Gene Symbol:
          LOC1720
          Gene Name:
          dihydrofolate reductase pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 82856803 - 82857769 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00983296_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC1720
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033423.1 1 787 123
          GenBank mRNA J00146.1 - 1 784 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200008200.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000493.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          reductase

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15261945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15261946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01319178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15261958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21401548 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3220872 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48564 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3220872 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P343095 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00329328 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3220872 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13293127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8043037 - -
          Affymetrix Human Gene 1.1 ST Array 8043036 - -
          Affymetrix HT HG-U133+ PM Array Plate 216659_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02007562.hg.1 TC02000493.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02007562.hg.1 NR_033423.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200156790.hg.1 ENST00000455595.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200156790.hg.1 NR_033423.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200156790.hg.1 ENST00000455595.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200156790.hg.1 NR_033423.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200156790.hg.1 TC0200008200.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200156791.hg.1 NR_033423.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200156791.hg.1 NR_033423.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200156791.hg.1 TC0200008200.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 216659_at - -
          Human Genome U133 Plus 2.0 Array 216659_PM_at - -
          Human Genome U133A 2.0 Array 216659_at - -
          Human Genome U133A 2.0 Array 216659_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw556302 - -
          Human Genome U95 Set 31479_f_at - -
          Human Genome U95 Set 31479_PM_f_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216659_at10 - -
          Human Genome U133 Set 216659_PM_at - -
          Human Genome U133 Set 216659_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015886 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015887 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015886 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015887 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline