| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019648.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019648.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019648.hg.1 |
TC11001569.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019647.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019647.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019647.hg.1 |
TC11001569.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019651.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019651.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019651.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019651.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019651.hg.1 |
TC11001569.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180307.hg.1 |
TC1100010591.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010591.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180302.hg.1 |
TC1100010591.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180303.hg.1 |
TC1100010591.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000525641.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000527757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
ENST00000530387.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
NR_038309.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180306.hg.1 |
TC1100010591.hg.1 |
Clariom D TC |
| Human Genome U95 Set |
42231_PM_at |
- |
- |
| Human Genome U95 Set |
42231_at |
- |
- |