Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00984442_m1
          See other F10 GE Assays ›
          Gene Symbol:
          F10
          Gene Name:
          coagulation factor X
          Gene Aliases:
          FX, FXA
          Chromosome Location:
          Chr.13: 113122799 - 113149529 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00984442_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000504.3 NP_000495.1 3 - 4 317 65
          NM_001312674.1 NP_001299603.1 3 - 4 317 65
          NM_001312675.1 NP_001299604.1 3 - 4 317 65
          GenBank mRNA AK310775.1 - 3 - 4 278 65
          AK313798.1 - 3 - 4 273 65
          BC040125.1 - 3 - 4 264 65
          BC046125.1 - 3 - 4 298 5723510 65
          CB158437.1 - 3 - 4 317 65
          CD013940.1 - 3 - 4 305 65
          K03194.1 - 3 - 4 226 65
          M22613.1 - 3 - 4 206 65
          M57285.1 - 3 - 4 260 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300008109.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000417.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F10

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          signal peptide processing
          ER to Golgi vesicle-mediated transport
          blood coagulation
          blood coagulation, intrinsic pathway
          blood coagulation, extrinsic pathway
          peptidyl-glutamic acid carboxylation
          positive regulation of cell migration
          positive regulation of protein kinase B signaling
          serine-type endopeptidase activity
          calcium ion binding
          protein binding
          phospholipid binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375551.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000375559.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000409306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000410083.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000477269.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000483537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000483537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000483537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000498455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000498455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000498455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000498455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 ENST00000498455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008109.hg.1 NM_000504.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 120934 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0