Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013669.hg.1 |
TC04000989.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013669.hg.1 |
ENST00000343470.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013669.hg.1 |
ENST00000452476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013669.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013669.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400167798.hg.1 |
TC0400009856.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000343470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000452476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000502917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
ENST00000513174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_001145725.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009856.hg.1 |
NM_017816.2 |
Refseq |