Search
Search

TMEM81| Gene Symbol: TMEM81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203376.1 | NP_976310.1 | 1 | 1155 | 71 | |
| GenBank mRNA | AK303432.1 | - | 1 | 1212 | 71 | |
| AY359081.1 | - | 1 | 1155 | 71 | ||
| BC061592.1 | - | 1 | 1098 | 6202253 | 71 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20807000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806992 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745492 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15165980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745502 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20807003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20807004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745474 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20807005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806983 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20806996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00745491 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P46690 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3462 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3463 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P46690 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 28853 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30916 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P10470818 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P46690 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P46690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806998 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745502 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20807005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00745484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20806990 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P46690 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745486 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745493 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745474 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745472 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745501 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745494 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745476 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745479 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745491 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745499 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745502 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745484 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745497 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745496 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745489 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745481 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00745498 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P46690 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745502 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745493 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745476 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745498 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00745484 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_203376_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7923708 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7923710 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7923709 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7923711 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7923707 | - | - |
| Affymetrix Human Genome U219 Array | 11726958_at | - | - |
| Affymetrix Human Genome U219 Array | 11726958_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057370.hg.1 | TC01003741.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057370.hg.1 | ENST00000367167.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057370.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057371.hg.1 | TC01003741.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057371.hg.1 | ENST00000367167.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057371.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229165.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229165.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229165.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229165.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229165.hg.1 | TC0100017058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229163.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229163.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229163.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229163.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229163.hg.1 | TC0100017058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017058.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229162.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229162.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229162.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229162.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229162.hg.1 | TC0100017058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229164.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229164.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229164.hg.1 | ENST00000367167.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229164.hg.1 | NM_203376.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229164.hg.1 | TC0100017058.hg.1 | Clariom D TC |
| Human Genome U95 Set | 80265_at | - | - |
| Human Genome U95 Set | 80265_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010175 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 214141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_42794617-S | NM_203376.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_42794617-S | AK303432.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_42794617-S | AY359081.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_42794617-S | BC061592.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_28556-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006590315 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006590315 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 130273 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_28556 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1725971 | BC061592.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1725971 | NM_203376.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1725971 | AK303432.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1725971 | AY359081.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1725971 | BC061592.1 | human_transcriptome |