| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030107.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030107.hg.1 |
ENST00000366592.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030107.hg.1 |
ENST00000391852.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030107.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030107.hg.1 |
TC01001950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030108.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030108.hg.1 |
ENST00000366592.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030108.hg.1 |
ENST00000391852.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030108.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030108.hg.1 |
TC01001950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030105.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030105.hg.1 |
ENST00000366592.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030105.hg.1 |
ENST00000391852.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030105.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030105.hg.1 |
TC01001950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030106.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030106.hg.1 |
ENST00000366592.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030106.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030106.hg.1 |
TC01001950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030110.hg.1 |
ENST00000366592.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030110.hg.1 |
ENST00000391852.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030110.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030110.hg.1 |
TC01001950.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188950.hg.1 |
TC0100012089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188951.hg.1 |
TC0100012089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
ENST00000366592.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
ENST00000454895.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
NM_003272.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188954.hg.1 |
TC0100012089.hg.1 |
Clariom D TC |