Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00991639_s1
          See other DKFZP434I0714 GE Assays ›
          Gene Symbol:
          DKFZP434I0714
          Gene Name:
          uncharacterized protein DKFZP434I0714
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 152536264 - 152539263 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00991639_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DKFZP434I0714
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033797.2 1 2058 131
          GenBank mRNA AL137273.1 - 1 2058 131
          BC034819.1 - 1 2029 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400009018.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000764.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23695565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23695558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23695555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03533870 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 180443 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 59289 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 385583 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 234692 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4195 - -
          Agilent Human Genome CGH Microarray 44B A_14_P138148 - -
          Agilent Human Genome CGH Microarray 44A A_14_P138148 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59316 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P413303 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P160254 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P138148 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P138148 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 113365 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 43442 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16910075 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P138148 - -
          Agilent Human 2 cDNA Microarray (G4101A) 999522 - -
          Agilent Human 2 cDNA Microarray (G4101A) 999522 - -
          Agilent Human 1B Microarray G4111A A_32_P477745 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P138148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8097812 - -
          Affymetrix Human Gene 1.1 ST Array 8097811 - -
          Affymetrix HT HG-U133+ PM Array Plate 231954_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000584019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400009018.hg.1 NR_039658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04010509.hg.1 TC04000764.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04010509.hg.1 NR_033797.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04010509.hg.1 NR_039658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04010508.hg.1 TC04000764.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04010508.hg.1 NR_033797.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04010510.hg.1 TC04000764.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04010510.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161841.hg.1 TC0400009018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161839.hg.1 TC0400009018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000584019.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 NR_039658.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000584019.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 NR_039658.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400161840.hg.1 TC0400009018.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000584019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000594836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 ENST00000604157.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 NR_033797.2 Refseq
          Affymetrix Human Clariom D Assay TC0400009018.hg.1 NR_039658.1 Refseq
          Human Genome U133 Plus 2.0 Array 231954_PM_at - -
          Human Genome U133 Plus 2.0 Array 231954_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231954_at4 - -
          Human Genome U95 Set 61838_at - -
          Human Genome U95 Set 61838_PM_at - -
          Human Genome U133 Set 231954_at - -
          Human Genome U133 Set 231954_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026163 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026164 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026163 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026164 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 159541 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1741621 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1741621 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1741621 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1741621 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741621 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0