| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013970.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013970.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013970.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013970.hg.1 |
TC22000826.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008828.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000216252.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000459687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
ENST00000491254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
NM_032758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163925.hg.1 |
TC2200008828.hg.1 |
Clariom D TC |