| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000392650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000410094.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000424778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000479356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000585283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
ENST00000585283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011653.hg.1 |
NM_001144952.1 |
Refseq |