Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00993250_g1
          See other LECT1 GE Assays ›
          Gene Symbol:
          LECT1
          Gene Name:
          leukocyte cell derived chemotaxin 1
          Gene Aliases:
          BRICD3, CHM-I, CHM1, MYETS1
          Chromosome Location:
          Chr.13: 52703264 - 52739812 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00993250_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LECT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011705.1 NP_001011705.1 1 - 2 184 68
          NM_007015.2 NP_008946.1 1 - 2 184 68
          XM_011534897.2 XP_011533199.1 1 - 2 182 68
          XM_011534898.2 XP_011533200.1 1 - 2 183 68
          XM_011534899.2 XP_011533201.1 1 - 2 184 68
          XM_011534900.2 XP_011533202.1 1 - 2 182 68
          GenBank mRNA AB006000.1 - 1 - 2 73 68
          BC025659.1 - 1 - 2 184 5198878 68
          CR541910.1 - 1 - 2 73 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009023.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000691.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LECT1

          Gene Ontology Categories:

          Function(s)

          skeletal system development
          proteoglycan metabolic process
          negative regulation of angiogenesis
          cell differentiation
          cartilage development

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13008129.hg.1 ENST00000377962.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008129.hg.1 ENST00000448904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008129.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008129.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008129.hg.1 TC13000691.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13008133.hg.1 ENST00000448904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008133.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008133.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008133.hg.1 TC13000691.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13008130.hg.1 ENST00000377962.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008130.hg.1 ENST00000448904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008130.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008130.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008130.hg.1 TC13000691.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13008132.hg.1 ENST00000377962.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008132.hg.1 ENST00000448904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008132.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008132.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008132.hg.1 TC13000691.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161069.hg.1 TC1300009023.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161070.hg.1 TC1300009023.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161067.hg.1 TC1300009023.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161071.hg.1 TC1300009023.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300161066.hg.1 TC1300009023.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000377962.7 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000431550.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 ENST00000448904.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_001011705.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq
          Affymetrix Human Clariom D Assay TC1300009023.hg.1 NM_007015.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline