Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00996913_m1
          See other PLA2G4A GE Assays ›
          Gene Symbol:
          PLA2G4A
          Gene Name:
          phospholipase A2 group IVA
          Gene Aliases:
          PLA2G4, cPLA2, cPLA2-alpha
          Chromosome Location:
          Chr.1: 186828900 - 186989896 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00996913_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLA2G4A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001311193.1 NP_001298122.1 13 - 14 1790 75
          NM_024420.2 NP_077734.1 15 - 16 1970 75
          XM_005245267.3 XP_005245324.2 13 - 14 1791 75
          XM_011509642.2 XP_011507944.1 15 - 16 1921 75
          GenBank mRNA AK290336.1 - 15 - 16 1921 75
          AK302938.1 - 13 - 14 1700 75
          BC114340.2 - 15 - 16 1867 40034995 75
          M68874.1 - 15 - 16 1903 75
          M72393.1 - 15 - 16 1890 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010939.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001607.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLA2G4A

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          luteolysis
          phospholipid metabolic process
          phosphatidic acid biosynthetic process
          platelet activating factor biosynthetic process
          icosanoid metabolic process
          aging
          positive regulation of cell proliferation
          phospholipid catabolic process
          response to heat
          response to lithium ion
          response to organonitrogen compound
          arachidonic acid metabolic process
          positive regulation of bone mineralization
          positive regulation of vesicle fusion
          positive regulation of prostaglandin biosynthetic process
          positive regulation of fever generation
          response to lipopolysaccharide
          response to vitamin D
          cardiolipin acyl-chain remodeling
          phosphatidylglycerol acyl-chain remodeling
          phosphatidylinositol acyl-chain remodeling
          phosphatidylserine acyl-chain remodeling
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          response to hydrogen peroxide
          positive regulation of apoptotic process
          surfactant homeostasis
          icosanoid biosynthetic process
          decidualization
          arachidonic acid secretion
          response to glucocorticoid
          response to calcium ion
          response to methylmercury
          cellular response to antibiotic
          lysophospholipase activity
          phospholipase A2 activity
          calcium ion binding
          calcium-dependent phospholipid binding
          phosphatidylcholine 1-acylhydrolase activity
          histone acetyltransferase binding
          calcium-dependent phospholipase A2 activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7sm7s:80/100.66.76.102:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0