Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00997374_m1
          See other SLC6A3 GE Assays ›
          Gene Symbol:
          SLC6A3
          Gene Name:
          solute carrier family 6 member 3
          Gene Aliases:
          DAT, DAT1, PKDYS, PKDYS1
          Chromosome Location:
          Chr.5: 1392794 - 1445440 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00997374_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC6A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044.5 NP_001035.1 9 - 10 1405 56
          GenBank mRNA BC132977.1 - 8 - 9 1318 56
          BC133003.1 - 8 - 9 1318 56
          EF174603.1 - 9 - 10 1395 56
          L24178.1 - 8 - 9 1286 56
          M95167.1 - 9 - 10 1395 56
          S44626.1 - 8 - 9 1292 56
          S46955.1 - 9 - 10 1368 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500009861.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001109.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC6A3

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          neurotransmitter transport
          amino acid transport
          lactation
          sensory perception of smell
          locomotory behavior
          response to xenobiotic stimulus
          response to iron ion
          monoamine transport
          dopamine transport
          adenohypophysis development
          response to nicotine
          sodium ion transmembrane transport
          positive regulation of multicellular organism growth
          regulation of dopamine metabolic process
          response to cocaine
          dopamine biosynthetic process
          dopamine catabolic process
          response to ethanol
          cognition
          dopamine uptake involved in synaptic transmission
          response to cAMP
          norepinephrine uptake
          prepulse inhibition
          dopamine uptake
          hyaloid vascular plexus regression
          protease binding
          receptor binding
          neurotransmitter transporter activity
          dopamine:sodium symporter activity
          norepinephrine:sodium symporter activity
          protein binding
          monoamine transmembrane transporter activity
          symporter activity
          transmembrane transporter activity
          dopamine binding
          amine binding
          macromolecular complex binding
          metal ion binding
          protein phosphatase 2A binding
          heterocyclic compound binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05016158.hg.1 ENST00000270349.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016158.hg.1 ENST00000453492.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016158.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016158.hg.1 TC05001109.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016157.hg.1 ENST00000270349.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016157.hg.1 ENST00000453492.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016157.hg.1 NM_001044.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016157.hg.1 TC05001109.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500169620.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169620.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500169620.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169620.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500169620.hg.1 TC0500009861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000511750.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 ENST00000512002.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay TC0500009861.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500169619.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169619.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500169619.hg.1 ENST00000270349.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169619.hg.1 NM_001044.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500169619.hg.1 TC0500009861.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline