| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012915.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012915.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012915.hg.1 |
TC01000811.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012917.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012917.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012917.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012917.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012917.hg.1 |
TC01000811.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100163389.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163389.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163389.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163389.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163389.hg.1 |
TC0100008882.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008882.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
ENST00000370662.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
ENST00000370665.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
NM_021233.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
NM_058248.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100163391.hg.1 |
TC0100008882.hg.1 |
Clariom D TC |