Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167556.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167556.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167556.hg.1 |
TC1900008649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167556.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167556.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167555.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167555.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167555.hg.1 |
TC1900008649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167555.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167555.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
ENST00000396477.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008649.hg.1 |
NM_001278392.1 |
Refseq |