| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804612 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369341 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804651 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804590 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804629 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804571 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804602 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804608 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804588 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804658 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369326 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804574 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804585 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369339 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804641 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804614 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369355 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804601 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804633 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369352 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369321 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369316 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369353 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804594 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804632 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804598 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804591 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804628 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369323 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369351 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804587 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804613 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804642 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804639 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804572 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369365 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804617 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804616 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804611 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804580 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804655 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804577 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804659 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804640 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804586 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804610 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16561743 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804618 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369324 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804595 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804603 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804605 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804581 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804593 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804582 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804645 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804657 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369302 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804660 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369340 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804596 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08369368 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804630 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804653 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804643 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28804631 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
154226 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3248987 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0716 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0717 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
40124 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P19551797 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3248987 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3248987 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804586 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804613 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804596 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804594 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804580 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804590 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804614 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08369323 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804591 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804584 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804581 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804617 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804588 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804608 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804605 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804587 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804616 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08369316 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804602 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804582 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804610 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804611 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804592 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804591 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804586 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804596 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08369323 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804587 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804614 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804602 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08369316 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804588 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804610 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804608 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804581 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804613 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804590 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804605 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804617 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804582 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804592 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804611 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804616 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804594 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804580 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28804584 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P19551797 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19551797 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
127651 |
NM_001080502.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
127651 |
XM_495909.2 |
refseq_mrna_human_XM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
40518 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19551797 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369350 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369368 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561745 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369345 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369317 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369331 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369348 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369319 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369358 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369314 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369312 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369340 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369376 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369333 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369321 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369328 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369338 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369325 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369362 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369299 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369307 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369372 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369309 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369355 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369342 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561741 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369335 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369370 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561756 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369336 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561747 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561752 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369375 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369365 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369360 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16561754 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369305 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369353 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369303 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369323 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08369367 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19551797 |
- |
- |