Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01001913_m1
          See other NCOA5 GE Assays ›
          Gene Symbol:
          NCOA5
          Gene Name:
          nuclear receptor coactivator 5
          Gene Aliases:
          CIA, bA465L10.6
          Chromosome Location:
          Chr.20: 46060985 - 46089962 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01001913_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NCOA5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020967.2 NP_066018.1 5 - 6 775 64
          XM_005260474.3 XP_005260531.1 4 - 5 729 64
          XM_011528951.2 XP_011527253.1 5 - 6 790 64
          XM_017027988.1 XP_016883477.1 4 - 5 729 64
          GenBank mRNA AB046857.1 - 3 - 4 323 64
          AF230533.1 - 5 - 6 747 64
          AF470686.1 - 4 - 5 1151 64
          BC140836.1 - 4 - 5 664 9021704 64
          BC151133.1 - 4 - 5 664 9057033 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009259.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000896.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NCOA5

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          glucose homeostasis
          negative regulation of insulin receptor signaling pathway
          chromatin binding
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20012253.hg.1 ENST00000290231.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20012253.hg.1 NM_020967.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20012253.hg.1 TC20000896.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000164828.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164828.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000164828.hg.1 TC2000009259.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000164828.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164828.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000290231.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 ENST00000372291.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009259.hg.1 NM_020967.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 166888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline