| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011835.hg.1 |
ENST00000290913.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011835.hg.1 |
ENST00000508789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011835.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011835.hg.1 |
TC03000660.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011834.hg.1 |
ENST00000290913.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011834.hg.1 |
ENST00000508789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011834.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011834.hg.1 |
TC03000660.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300161649.hg.1 |
TC0300008686.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |